Order Entry
Czech Republic
ContactUsLinkComponent
4-(Dimethoxymethyl)aniline 95%
4-(Dimethoxymethyl)aniline 95%
Catalog # APOSOR55356-5G
Supplier:  Apollo Scientific
CAS Number:  
4-(Dimethoxymethyl)aniline 95%
Catalog # APOSOR55356-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :167.205 g/mol
Storage Temperature:Ambient
CAS Number:111616-46-7
Formula:C₉H₁₃NO₂
MDL Number:MFCD20542810