Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
22-(tert-Butoxy)-22-oxodocosanoic acid
Catalog # APOSOR55180-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
22-(tert-Butoxy)-22-oxodocosanoic acid
Catalog # APOSOR55180-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :426.673 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1642333-05-8
Formula:C₂₆H₅₀O₄
MDL Number:MFCD30830215