
Methyl 5-amino-2-bromobenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6942-37-6
:
Formula:C₈H₈BrNO₂
MDL Number:MFCD00085568
:230.059 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
Frequently Bought Together