
4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14160-91-9
:
Formula:C₆H₄Cl₂N₂O
MDL Number:MFCD09909782
:191.015 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together