Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 3-(4-methoxyphenyl)propanoate 98%
Irritant
Ethyl 3-(4-methoxyphenyl)propanoate 98%
Catalog # APOSOR53089-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 3-(4-methoxyphenyl)propanoate 98%
Catalog # APOSOR53089-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.254 g/mol
Melting Pt:52 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:22767-72-2
Formula:C₁₂H₁₆O₃
MDL Number:MFCD02258632