
6-Chloro-1H-pyrazolo[3,4-b]pyridin-3-amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1352909-30-8
:
Formula:C₆H₅ClN₄
MDL Number:MFCD20492485
:168.584 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together