Order Entry
Czech Republic
ContactUsLinkComponent
N,2-Dimethoxy-N-methylacetamide 95%
Irritant
N,2-Dimethoxy-N-methylacetamide 95%
Catalog # APOSOR52620-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N,2-Dimethoxy-N-methylacetamide 95%
Catalog # APOSOR52620-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :133.146 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:132289-57-7
Formula:C₅H₁₁NO₃
MDL Number:MFCD11848891