
4-Chloro-6-methyl-1H-pyrazolo[3,4-d]pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:30129-53-4
:
Formula:C₆H₅ClN₄
MDL Number:MFCD10697160
:168.584 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together