Order Entry
Czech Republic
ContactUsLinkComponent
2-Azaspiro[3.3]heptan-6-ol hydrochloride 97%
Irritant
2-Azaspiro[3.3]heptan-6-ol hydrochloride 97%
Catalog # APOSOR52371-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Azaspiro[3.3]heptan-6-ol hydrochloride 97%
Catalog # APOSOR52371-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :149.619 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1630907-10-6
Formula:C₆H₁₁NO·ClH
MDL Number:MFCD23703064