
2-(Prop-1-en-2-yl)aniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52562-19-3
:
Formula:C₉H₁₁N
MDL Number:MFCD00007719
:133.19 g/mol
Boiling Pt:95 °C
Flash Pt:78 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together