(S)-1-(3-Bromo-5-chloropyridin-2-yl)ethanamine
Catalog # APOSOR52304-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:235.509 g/mol
Storage Temperature:Ambient
CAS Number:1259882-26-2
Formula:C₇H₈BrClN₂
MDL Number:MFCD18209621