(S)-1-(2,6-Dichloropyridin-3-yl)ethanamine
Catalog # APOSOR52301-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:191.058 g/mol
Storage Temperature:Ambient
CAS Number:1213971-04-0
Formula:C₇H₈Cl₂N₂
MDL Number:MFCD09830322