Order Entry
Czech Republic
ContactUsLinkComponent
(S)-1-(5-Methylpyrimidin-2-yl)ethanamine
(S)-1-(5-Methylpyrimidin-2-yl)ethanamine
Catalog # APOSOR52299-500MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-1-(5-Methylpyrimidin-2-yl)ethanamine
Catalog # APOSOR52299-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :137.182 g/mol
Storage Temperature:Ambient
CAS Number:1568163-63-2
Formula:C₇H₁₁N₃
MDL Number:MFCD24249248