(S)-1-(Furo[2,3-b]pyridin-5-yl)ethanamine
Catalog # APOSOR52298-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:162.189 g/mol
Storage Temperature:Ambient
CAS Number:1932107-45-3
Formula:C₉H₁₀N₂O
MDL Number:MFCD29042488