Order Entry
Czech Republic
ContactUsLinkComponent
 
(S)-1-(Isoquinolin-8-yl)ethanamine
Catalog # APOSOR52296-500MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-1-(Isoquinolin-8-yl)ethanamine
Catalog # APOSOR52296-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :172.226 g/mol
Storage Temperature:Ambient
CAS Number:1344594-30-4
Formula:C₁₁H₁₂N₂
MDL Number:MFCD21348027