(S)-1-(Isoquinolin-6-yl)ethanamine
Catalog # APOSOR52295-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:172.226 g/mol
Storage Temperature:Ambient
CAS Number:1344588-21-1
Formula:C₁₁H₁₂N₂
MDL Number:MFCD21348045