(S)-1-(1H-Indazol-5-yl)ethanamine
Catalog # APOSOR52291-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:161.204 g/mol
Storage Temperature:Ambient
CAS Number:1259610-88-2
Formula:C₉H₁₁N₃
MDL Number:MFCD18209561