
2,2,6,6-Tetramethyloxan-4-one
Catalog # APOSOR52276-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:156.222 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1197-66-6
Formula:C₉H₁₆O₂
MDL Number:MFCD12828096