1-Propylpiperazine hydrochloride
Catalog # APOSOR51746-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:164.676 g/mol
Storage Temperature:Ambient
CAS Number:71888-55-6
Formula:C₇H₁₆N₂·ClH
MDL Number:MFCD11575790