tert-Butyl 3-bromo-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥98%
Catalog # APOSOR510250-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:302.168 g/mol
Storage Temperature:Ambient
CAS Number:1936429-06-9
Formula:C₁₁H₁₆BrN₃O₂
MDL Number:MFCD30751004