Order Entry
Czech Republic
ContactUsLinkComponent
1,5-Dichloro-2-methoxy-3-nitrobenzene ≥98%
1,5-Dichloro-2-methoxy-3-nitrobenzene ≥98%
Catalog # APOSOR510248-250MG
Supplier:  Apollo Scientific
CAS Number:  
1,5-Dichloro-2-methoxy-3-nitrobenzene ≥98%
Catalog # APOSOR510248-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :222.026 g/mol
Storage Temperature:Ambient
CAS Number:37138-82-2
Formula:C₇H₅Cl₂NO₃
MDL Number:MFCD09037656