Order Entry
Czech Republic
ContactUsLinkComponent
2-Chlorodibenzo[b,f]oxepine ≥98%
2-Chlorodibenzo[b,f]oxepine ≥98%
Catalog # APOSOR510239-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chlorodibenzo[b,f]oxepine ≥98%
Catalog # APOSOR510239-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.674 g/mol
Storage Temperature:Ambient
CAS Number:25558-88-7
Formula:C₁₄H₉ClO
MDL Number:MFCD22124643