7-Bromo-3-chloro-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%
Catalog # APOSOR510233-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.487 g/mol
Storage Temperature:Ambient
CAS Number:1785021-08-0
Formula:C₈H₄BrClN₂O