Order Entry
Czech Republic
ContactUsLinkComponent
1-(7-Bromobenzo[d][1,3]dioxol-4-yl)ethan-1-one ≥98%
1-(7-Bromobenzo[d][1,3]dioxol-4-yl)ethan-1-one ≥98%
Catalog # APOSOR510231-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(7-Bromobenzo[d][1,3]dioxol-4-yl)ethan-1-one ≥98%
Catalog # APOSOR510231-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.054 g/mol
Storage Temperature:Ambient
CAS Number:1892297-27-6
Formula:C₉H₇BrO₃
MDL Number:MFCD30738025