Methyl 4-chloro-3-cyanobenzoate ≥98%
Catalog # APOSOR510225-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:195.602 g/mol
Storage Temperature:Ambient
CAS Number:181282-80-4
Formula:C₉H₆ClNO₂
MDL Number:MFCD18398898