Order Entry
Czech Republic
ContactUsLinkComponent
4-Hydroxy-7-methoxy-6-nitroquinoline-3-carbonitrile ≥98%
4-Hydroxy-7-methoxy-6-nitroquinoline-3-carbonitrile ≥98%
Catalog # APOSOR510212-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Hydroxy-7-methoxy-6-nitroquinoline-3-carbonitrile ≥98%
Catalog # APOSOR510212-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :245.191 g/mol
Storage Temperature:Ambient
CAS Number:492456-52-7
Formula:C₁₁H₇N₃O₄
MDL Number:MFCD13181118