Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-1,2-dichloro-3-nitrobenzene ≥98%
5-Bromo-1,2-dichloro-3-nitrobenzene ≥98%
Catalog # APOSOR510209-100MG
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-1,2-dichloro-3-nitrobenzene ≥98%
Catalog # APOSOR510209-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :270.896 g/mol
Storage Temperature:Ambient
CAS Number:1805596-72-8
Formula:C₆H₂BrCl₂NO₂
MDL Number:MFCD28250219