Order Entry
Czech Republic
ContactUsLinkComponent
3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde ≥98%
3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde ≥98%
Catalog # APOSOR510191-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole-4-carbaldehyde ≥98%
Catalog # APOSOR510191-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :306.1 g/mol
Storage Temperature:Ambient
CAS Number:1627924-19-9
Formula:C₉H₁₁IN₂O₂