6-Chloro-2-morpholinonicotinic acid ≥98%
Catalog # APOSOR510187-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:242.659 g/mol
Storage Temperature:Ambient
CAS Number:305863-07-4
Formula:C₁₀H₁₁ClN₂O₃
MDL Number:MFCD00452940