4,5,6,7-Tetrahydro-1H-indazole-5-carboxylic acid ≥98%
Catalog # APOSOR510180-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:166.177 g/mol
Storage Temperature:Ambient
CAS Number:52834-38-5
Formula:C₈H₁₀N₂O₂
MDL Number:MFCD20647600