Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 5-bromo-4-methyl-1H-indole-2-carboxylate ≥98%
Methyl 5-bromo-4-methyl-1H-indole-2-carboxylate ≥98%
Catalog # APOSOR510178-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 5-bromo-4-methyl-1H-indole-2-carboxylate ≥98%
Catalog # APOSOR510178-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :268.107 g/mol
Storage Temperature:Ambient
CAS Number:1857296-39-9
Formula:C₁₁H₁₀BrNO₂
MDL Number:MFCD29035115