Order Entry
Czech Republic
ContactUsLinkComponent
 
7-Bromochromane-3-carboxylic acid ≥98%
Catalog # APOSOR510167-100MG
Supplier:  Apollo Scientific
CAS Number:  
7-Bromochromane-3-carboxylic acid ≥98%
Catalog # APOSOR510167-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :257.081 g/mol
Storage Temperature:Ambient
CAS Number:1781758-38-0
Formula:C₁₀H₉BrO₃
MDL Number:MFCD27922386