Order Entry
Czech Republic
ContactUsLinkComponent
(7-Chloronaphthalen-1-yl)methanamine hydrochloride ≥98%
(7-Chloronaphthalen-1-yl)methanamine hydrochloride ≥98%
Catalog # APOSOR510161-100MG
Supplier:  Apollo Scientific
CAS Number:  
(7-Chloronaphthalen-1-yl)methanamine hydrochloride ≥98%
Catalog # APOSOR510161-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :228.118 g/mol
Storage Temperature:Ambient
CAS Number:1093651-77-4
Formula:C₁₁H₁₀ClN·ClH
MDL Number:MFCD31617675