1-Bromo-2,6-naphthyridin-3-amine ≥98%
Catalog # APOSOR510160-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:224.057 g/mol
Storage Temperature:Ambient
CAS Number:3611-11-8
Formula:C₈H₆BrN₃
MDL Number:MFCD00160697