7-Bromonaphthalen-1-amine ≥98%
Catalog # APOSOR510159-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:222.081 g/mol
Storage Temperature:Ambient
CAS Number:136924-78-2
Formula:C₁₀H₈BrN
MDL Number:MFCD17012507