Order Entry
Czech Republic
ContactUsLinkComponent
(3-Chlorothiophen-2-yl)methanamine hydrochloride ≥98%
(3-Chlorothiophen-2-yl)methanamine hydrochloride ≥98%
Catalog # APOSOR510148-100MG
Supplier:  Apollo Scientific
CAS Number:  
(3-Chlorothiophen-2-yl)methanamine hydrochloride ≥98%
Catalog # APOSOR510148-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :184.087 g/mol
Storage Temperature:Ambient
CAS Number:643088-03-3
Formula:C₅H₆ClNS·ClH