Order Entry
Czech Republic
ContactUsLinkComponent
(5-Chloro-1H-indol-2-yl)methanol ≥98%
(5-Chloro-1H-indol-2-yl)methanol ≥98%
Catalog # APOSOR510147-100MG
Supplier:  Apollo Scientific
CAS Number:  
(5-Chloro-1H-indol-2-yl)methanol ≥98%
Catalog # APOSOR510147-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :181.619 g/mol
Storage Temperature:Ambient
CAS Number:53590-47-9
Formula:C₉H₈ClNO
MDL Number:MFCD19301593