Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromooxazole-4-carbonitrile ≥98%
2-Bromooxazole-4-carbonitrile ≥98%
Catalog # APOSOR510141-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-Bromooxazole-4-carbonitrile ≥98%
Catalog # APOSOR510141-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :172.968 g/mol
Storage Temperature:Ambient
CAS Number:1240608-82-5
Formula:C₄HBrN₂O