Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-6-methylpyridine-4-carbonitrile ≥98%
2-Chloro-6-methylpyridine-4-carbonitrile ≥98%
Catalog # APOSOR510133-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-methylpyridine-4-carbonitrile ≥98%
Catalog # APOSOR510133-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :152.581 g/mol
Storage Temperature:Ambient
CAS Number:25462-98-0
Formula:C₇H₅ClN₂
MDL Number:MFCD16658610