Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Methyl-1H-pyrrol-3-yl)ethanone ≥98%
1-(4-Methyl-1H-pyrrol-3-yl)ethanone ≥98%
Catalog # APOSOR510125-100MG
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Methyl-1H-pyrrol-3-yl)ethanone ≥98%
Catalog # APOSOR510125-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :123.152 g/mol
Storage Temperature:Ambient
CAS Number:18818-30-9
Formula:C₇H₉NO
MDL Number:MFCD00111582