Order Entry
Czech Republic
ContactUsLinkComponent
2-Iodo-6-methoxy-3-nitropyridine ≥98%
2-Iodo-6-methoxy-3-nitropyridine ≥98%
Catalog # APOSOR510113-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Iodo-6-methoxy-3-nitropyridine ≥98%
Catalog # APOSOR510113-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :280.02 g/mol
Storage Temperature:Refrigerator
CAS Number:1600094-08-3
Formula:C₆H₅IN₂O₃
MDL Number:MFCD30241102