Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 4-bromo-3-(hydroxymethyl)benzoate ≥95%
Methyl 4-bromo-3-(hydroxymethyl)benzoate ≥95%
Catalog # APOSOR510049-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-bromo-3-(hydroxymethyl)benzoate ≥95%
Catalog # APOSOR510049-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :245.07 g/mol
Storage Temperature:Ambient
CAS Number:254746-40-2
Formula:C₉H₉BrO₃
MDL Number:MFCD15527255