4-Bromo-2-methyl-2H-pyrazolo[3,4-c]pyridine ≥97%
Catalog # APOSOR510031-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:212.047 g/mol
Storage Temperature:Refrigerator
CAS Number:2177257-61-1
Formula:C₇H₆BrN₃
MDL Number:MFCD31652709