Order Entry
Czech Republic
ContactUsLinkComponent
4,5,6,7-Tetrahydro-1H-indazol-5-amine dihydrochloride ≥96%
4,5,6,7-Tetrahydro-1H-indazol-5-amine dihydrochloride ≥96%
Catalog # APOSOR510030-250MG
Supplier:  Apollo Scientific
CAS Number:  
4,5,6,7-Tetrahydro-1H-indazol-5-amine dihydrochloride ≥96%
Catalog # APOSOR510030-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :210.104 g/mol
Storage Temperature:Refrigerator
CAS Number:74197-17-4
Formula:C₇H₁₁N₃·2ClH
MDL Number:MFCD09028167