Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Formyl-4-methyl-1H-indole-2-carbonitrile
5-Formyl-4-methyl-1H-indole-2-carbonitrile ≥97%
5-Formyl-4-methyl-1H-indole-2-carbonitrile ≥97%
APOSOR510022-250MG
:
Apollo Scientific
:
5-Formyl-4-methyl-1H-indole-2-carbonitrile ≥97%
APOSOR510022-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
184.194 g/mol
:
Refrigerator
:
1857296-22-0
:
C₁₁H₈N₂O
:
MFCD31567509