Order Entry
Czech Republic
ContactUsLinkComponent
5-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde ≥97%
5-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde ≥97%
Catalog # APOSOR510017-250MG
Supplier:  Apollo Scientific
CAS Number:  
5-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde ≥97%
Catalog # APOSOR510017-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :180.591 g/mol
Storage Temperature:Refrigerator
CAS Number:198895-50-0
Formula:C₈H₅ClN₂O