6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine ≥97%
Catalog # APOSOR510011-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:167.596 g/mol
Storage Temperature:Refrigerator
CAS Number:1558302-68-3
Formula:C₇H₆ClN₃
MDL Number:MFCD28608322