(6-Methylpyridin-2-yl)methanamine hydrochloride ≥98%
Catalog # APOSOR510008-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:158.629 g/mol
Storage Temperature:Ambient
CAS Number:1365836-53-8
Formula:C₇H₁₀N₂·ClH
MDL Number:MFCD09878926