4-(2,2-Dimethylpropanoylamino)benzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:438191-85-6
:
Formula:C₁₁H₁₄ClNO₃S
MDL Number:MFCD09971327
:275.752 g/mol
Storage Temperature:Refrigerator
UN:1759
ADR:8,III
Frequently Bought Together