Ethyl 5-chloropyrazolo[1,5-a]pyrimidine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1224944-77-7
:
Formula:C₉H₈ClN₃O₂
MDL Number:MFCD12407819
:225.632 g/mol
Storage Temperature:Ambient
Frequently Bought Together